Learn More
Scientific Gear LLC The MKV-710S is a compact volumetric KF titrator with a large touch screen, which can measure in the range of 1 ppm to 100% H2O precisely in the sample of petroleum, chemical, and food industry.
SDP
Supplier:  Scientific Gear LLC MKV710S
Encompass_Preferred
Request Bulk Order
The MKV-710S is a compact volumetric Karl fischer titrator with a large touch screen, which can measure in the range of 1 ppm to 100% H2O precisely in the sample of petroleum, chemical, and food industry. The equipped 10mL burette discharge precision is ±0.015mL and Repeatability is ±0.005mL. Required solvent volume is 30 to 100mL. Measurement data can be saved for up to 100. The RS-232C and USB interface are equipped to connect an optional printer, electronic balance, and optional data acquisition software (SOFT-CAP).
Catalog No. NC2129975