Learn More
Scientific Gear LLC Auto Sampler AS 8030 can perform multiple sample measurements continuously including rinsing and drying the measuring cell and tubes in conjunction with KEM DA-860, DA-850, DA-840 series.
SDP
Supplier:  Scientific Gear LLC AS8030
Encompass_Preferred
Auto Sampler AS 8030 can perform multiple sample measurements continuously including rinsing and drying the measuring cell and tubes in conjunction with KEM DA-860, DA-850, and DA-840 series. In addition, we improved the drying time by using a high-volume drying pump and also enhanced the rinsing performance by using 2 kinds of rinse liquids. Moreover, the instrument has inherited the pressurized measurement function that returns the bubbles generated during sampling such as gasoline into the sample solution.With these improvements, the AS-8030 autosampler can provide operators with a convenient and highly reliable fully automatic measurement system. The number of sample positions is 30 and the size of sample vials is 20mL.
Catalog No. NC2129972