Learn More
Sklar Instruments Forceps, Surgi-OR Kelly Hemostatic Forceps, Sklar 5-1/2 in., Curved, Serrated, Stainless Steel, EA.
Supplier: Sklar Instruments 95442
Forceps, Surgi-OR Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Stainless Steel, EA. Surgi-OR Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Surgi-OR products are mid-grade instruments suitable for the operating room and other surgical centers. This product is curved with serrated tips and a length of 5-1/2 inches. Lifetime warranty against all manufacturer defects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.