Learn More
Enzo Life Sciences Irisin (human), (recombinant) (active) (10µg)
Supplier: Enzo Life Sciences ADI9083070010
First to market active Irisin. Alternative name: FNDC5, FRCP2. Endotoxin: <0.01EU/µg. Purity: ≥90% (SDS-PAGE). MW: ~25kDa. Formulation: Liquid. In 1X PBS. Source: Produced in CHO cells. aa 32-143, also known as Irisin, comprises the majority of the extracellular domain of FNDC5. It is fused at the N-terminus to a FLAG-tag with an 8 aa linker between the FLAG-tag and Irisin. Long Term Storage: -80°C. Use/Stability: Stable for up to a week when stored at +4°C and at least 6 months when stored at -80°C. Handling: Avoid freeze/thaw cycles. Applications: ELISA, WB SDS-PAGE, Activity assay.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.