Learn More
Enzo Life Sciences SCREEN-WELL Ion Channel ligand library (500 µl/well)
Supplier: Enzo Life Sciences BML28050500
The SCREEN-WELL Ion Channel Ligand Library contains 70 ion channel blockers and openers for use in characterizing and identifying ion channels in individual cells or tissue. Long Term Storage: -80°C. Applications: HTS .