Learn More
3M™ Intrinsically Safe Task Light
3M™ Hard Hat 150 Lumen, Intrinsically Safe Task Light is a versatile headlamp that allows hands free lighting for industrial workers who need Intrinsically safe certification.
Supplier: 3M™ 7010386282
Description
- Intrinsically Safe
- 150 Lumens
- IPX-7 Water resistant
- 5 modes
- Spot light and flood light
- Comes with 3M™ Dual Lock fasteners eliminating the need for a headband that could interfere with earmuffs and other attachments
- Comes with a rubber headband which provides a secure attachment to most 3M hard hats
- Comes with nylon headband which allows the lamp to be worn by the user without wearing a hard hat
- Versatile headlamp that allows hands free lighting for any industrial worker that needs intrinsically safe certification
- Comes with 3 AAA batteries
Specifications
3 AAA Batteries, 3M™ Dual Lock fasteners , Rubber headband, Nylon headband | |
HL300IS | |
Task Light | |
10/Cs. |