Learn More
BioMed Diagnostics A chromogenic medium for isolating pure cultures of vancomycin-resistant Enterococcus spp. Not for prevention, control or diagnosis. Sub-culture necessary for identification. | Available in: 20 pack
Supplier:  BioMed Diagnostics 11153002
Encompass_Preferred
COLOREX VRE prepared plated culture media is a selective and differential chromogenic medium, containing vancomycin, intended for use in isolation of pure cultures of vancomycin resistant Enterococcus faecium and Enterococcus faecalis (VRE) from laboratory samples. Further, colony color is used to differentiate transmissible forms of resistance (i.e., VanA & VanB resistance) from intrinsically resistant (i.e. VanC, VanD, VanE, etc.) strains. | Detects: vancomycin-resistant Enterococcus spp | Time to Result: Incubate at 37 2C (95-102F) for 24 hours under ambient atmosphere. | Available in: 20 pack
Catalog No. NC2582933