Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OraSure Technologies InteliSwab™ COVID-19 Rapid Test, Rx
This COVID-19 prescription rapid antigen test provides results in minutes with a unique all-in-one test device and nasal swab. InteliSwab™ COVID-19 Rapid Test Rx is one of the easiest rapid tests with less than 1 minute of hands on time and only 3 key steps: Swab, Swirl and See.
Supplier: OraSure Technologies 10010620
This item is not returnable.
View return policy
Description
- Self-collected, visual read lateral flow test
- Intuitive workflow: just swab, swirl, and see
- Workflow requires < 1 minute of active, hands-on time; 30-minute time to result
- Gentle lower nasal swab
- Overall test performance: Positive Percent Agreement 84%, Negative Percent Agreement 98%
- No precise number of drops required
Specifications
No | |
SARS-CoV-2 Nucleocapsid Protein Antigen | |
84% |
Waived (Prescription Home Use) | |
Anterior Nasal Swab | |
COVID-19 Rapid Antigen Test |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction