Learn More
Bevco Precision Manufacturing Co Integra Desk Height ISO 5 Cleanroom Black Vinyl Chair w/Arms; Medium Back; Articulating Seat & Back Tilt; Polished Aluminum Base; Interval Braking Casters
Supplier:  Bevco Precision Manufacturing Co 9051MC2BLKIBCSA5
Encompass_Preferred
BEVCO’s Integra ISO 5 Cleanroom Series is an upholstered chair that offers around-the-clock reliability for work environments which need to meet particulate goals of ISO 14644-1 . A unique filtration system traps particles for contamination-free comfort and has been tested by an independent laboratory to certify compliance to ISO 5. The Integra makes working easy!The medium backrest is specifically shaped to provide ample back support and comes standard with a heavy-duty polypropylene back shell to protect the upholstery.The generous seat provides an exceptional seating experience while the thick, resilient, high-density foam provides long-lasting comfort.The ergonomic adjustable arms help prevent neck & shoulder fatigue.This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations.
Catalog No. NC2130716