Learn More
Immunostics Hema-Screen SPECIFIC™ Mailing Envelope Box
Use this immunochemical fecal occult bood collection mailing envelope in conjunction with an iFOBT cassette and buffer tube to detect human hemoglobin [Hb (globin)] in stool.
Supplier: Immunostics HSGDENV20
This item is not returnable.
View return policy
Description
- 20 postal approved envelopes each containing 1 Devel-A-Tab™ slide, 2 collection tissues, 2 applicators, and patient instructions
- Can also be used as replacement envelopes for Hema-Screen SPECIFIC™ iFOBT Kit (23-040-035)
Intended for professional and laboratory use only
Specifications
Waived | |
Detection of Human hemoglobin in feces | |
20 Pack |
Mailing Envelopes | |
20 Mailing envelopes containing 1 DEVEL-A-TAB™ slide, 2 collection tissues, 2 applicator sticks, patient instructions, 1 product Insert |