Learn More
Enzo Life Sciences IHC antigen retrieval reagent (citrate, pH 8.0) (500ml)
SDP
Supplier:  Enzo Life Sciences ADI9502720500
Encompass_Preferred
IHC antigen retrieval reagent (citrate, pH 8.0) is used to recover antigens masked by because fixation in cross linking fixatives such as formalin. Formulation: Liquid. 10X citrate buffer, pH 8.0. Long Term Storage: Ambient. Applications: IHC .
Catalog No. 50-200-7123