Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2 Human, Isotype Control, Novus Biologicals™
Human Monoclonal Isotype Control
Supplier: Novus Biologicals NBP3068700.5MG
This item is not returnable.
View return policy
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for ControlSpecifications
IgG2 | |
MAB1641 | |
Lyophilized from a 0.2 um filtered solution in PBS. 0.5mg and 0.1 mg sizes are provided in liquid form, PBS | |
Human | |
Protein A purified | |
RUO | |
3501 | |
Human | |
Isotype Controls | |
IgG2 |
Monoclonal | |
Unconjugated | |
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
Human myeloma serum | |
0.5 mg | |
Immunology | |
Reconstitute with sterile 1 mL PBS | |
Store at 4°C in the dark. Do not freeze. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction