Learn More
Enzo Life Sciences IGF-I (human), (recombinant) (100µg)
Supplier: Enzo Life Sciences ADI9080590100
Alternative name: Insulin-like growth factor I. Purity: ≥95% (SDS and HPLC). MW: ~8kDa. Formulation: Lyophilized from a sterile filtered concentrated solution in PBS, pH 7.0. Source: Produced in E. coli. Long Term Storage: -20°C. Use/Stability: Lyophilized IGF-I although stable at room temperature for 3 weeks, should be stored desiccated below -18°C. Upon reconstitution IGF-I should be stored at +4°C between 2-7 days and for future use below -18°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Handling: Avoid freeze-thaw cycles. Applications: ELISA, WB Activity assay.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.