Learn More
Kamiya Biomedical IGA Reagent, 80mL+20mL/PK RX
Supplier:  Kamiya Biomedical KAI013
Encompass_Preferred
K-ASSAY IgA Assay Reagent. R1: 4 x 20mL (liquid), R2: 2 x 10mL (liquid). Assay Range: 20 - 800 mg/dL. The K-ASSAY IgG assay is intended for the quantitative determination of human IgA in serum by immunoturbidimetric assay. Measurement of IgA aids in the diagnosis of abnormal protein metabolism and the body's lack of ability to resist infectious agents. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Multi-Analyte Calibrator (Cat. No. KAI-016C). Recommended Controls: K-ASSAY Liquid ITA Control (Cat. No. K49C-2M or K49C-4M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
Catalog No. NC1944164
Due to product restrictions, please to purchase or view availability for this product.