Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Haemophilus influenzae type b; MinnA, Titered
Can be used to determine limit of detection (LOD), in diagnostic assay development or cross-reactivity studies
Supplier: ZeptoMetrix 0801680
Description
- Each frozen aliquot contains 1mL of pure, titered culture of Haemophilus influenzae
- Identification of the organism was confirmed by 16S sequencing
- Biosafety level 2
- Must be used within Biological Safety Level 2 facility or cabinet
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
2 | |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
ISO, 13485 | |
Solution | |
Titered Live Bacteria Control | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction