Learn More
Novus Biologicals™ Human C5b-9 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP266708
Description
The Human C5b-9 ELISA Kit (Colorimetric) measures human C5b-9 in serum, plasma and other biological fluids.Specifications
31.25 - 2000 ng/mL | |
This kit recognizes Human SC5b9 in samples. No significant cross-reactivity or interference between Human SC5b9 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
ELISA | |
Human | |
Storage of components varies. See protocol for specific instructions. | |
C5b-9 |
18.75 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human sC5b9 (soluble terminal complement complex C5b-9) in samples. No significant cross-reactivity or interference between Human sC5b9 and analogues was observed. | |
C5b-9 | |
1 Kit | |
MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex |