Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human BCMA/TNFRSF17 Quantikine ELISA Kit
$672.35 - $3832.94
Specifications
Assay Range | 31.30 to 2000 pg/mL |
---|---|
Assay Sensitivity | 10.8 pg/mL |
Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA), Heparin Plasma, Saliva, Urine |
Specificity | Natural and recombinant human BCMA. |
For Use With (Application) | ELISA |
Specifications
31.30 to 2000 pg/mL | |
Cell Culture Supernatant, Serum, Plasma (EDTA), Heparin Plasma, Saliva, Urine | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Human |
10.8 pg/mL | |
Natural and recombinant human BCMA. | |
608 | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title