Learn More
Tecan HMGB1 Express ELISA
Supplier:  Tecan 30164033
Encompass_Preferred
The new HMGB1 Express ELISA now allows to get results quicker as before as we were able to manage to reduce working time from 2 days to 4 hours. Furthermore, the new kit is more sensitive than the ST51011 HMGB1 ELISA due to newly used monoclonal antibody resulting in more endogenous HMGB1 to be detected by the HMGB1 Express ELISA. Still the kit correlates well to the predecessor HMGB1 ELISA
Catalog No. NC1927376