Learn More
Yamato Scientific America, Inc. High Purity DI System- 4-1/2" Diameter (10" Carbon, (2) 20" Mixed Bed, & 200 K DI Light). Type 2 and 3 water quality.
Supplier: Yamato Scientific America, Inc. WH501P
A pre-assembled cartridge system that provides deionized water using a staged filtration process. Water purification is provided using a three-stage process.