Learn More
World Precision Instrument High Intensity Fiber Optic Illumination Source-Z-LITE-186
Supplier: World Precision Instrument ZLITE186
The Z-LITE Fiber Optic Illuminator provides reliable, uninterrupted high-intensity light for microscopes. Z-LITE allows a continuous range of subdued or concentrated lighting controlled by a rotary dimmer on the front panel. Z-LITE may be used with a ring light and single or bifurcated flexible fiber bundles, enabling the light beam to be placed exactly where it is needed. Forced air cooling prolongs the lamp life. Lamp color temperature is 3350°K. An interlock switch automatically cuts off power when front panel is opened to replace bulb.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.