Learn More
Bellco Glass, Inc. The Bellco High Torque Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large v
Supplier: Bellco Glass, Inc. 777430230
The Bellco High Torque Digital Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large v