Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Monoclonal Antibody (2-2.2.14), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific 26183D488
Description
26183-D488 has been successfully used for immunofluorescence. DyLight 488 has an excitation/emission of 493/518 nm. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Immunocytochemistry | |
2-2.2.14 | |
DyLight 488 | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
50 μL | |
Primary | |
4° C, do not freeze | |
Liquid | |
Tag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction