Learn More
Sklar Instruments Curette, Gracey Curette, Sklar #1 / 2, Double End, Round Handle, Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 492635
Encompass_Preferred
Curette, Gracey Curette, Sklar, #1 / 2, Double End, Round Handle, Sharp, Straight, Smooth, Stainless Steel, EA. Sklar's Gracey Periodontal Curette is a double-ended dental curette used to scale and remove deposits from posterior teeth. The blade variations are specific to the particular posterior tooth surface being cleaned. This product is the size #1 / #2 curette. It has two sharp, angled, hook style tips and a round handle. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2371