Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Gooseneck Lamp
Flexible, inexpensive lighting for many tasks in your lab.
Supplier: American Educational Products 7S42211
Includes:
- 12 in. (30cm) gooseneck
- Cast-iron base
- 6 ft. (15m) cord
Description
Traditional UL-listed, black gooseneck lamp is perfect for a wide range of lab illumination tasks. Use for fill light in microvideo applications, to light dissections and for photomicroscopy.
Specifications
Gooseneck Lamp | |
UL | |
Fill light in microvideo application, light dissections, photomicroscopy | |
12 in. Gooseneck, cast Iron base, 6 ft. cord, two wire cord |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction