Learn More
Perkin Elmer US LLC Glow Plug for External Ignite FID
Supplier: Perkin Elmer US LLC 00091279
Two sizes of aluminum capsules are available. The smaller, with an internal volume of 4 mL, can hold sample weights up to 2.5 mg. The larger, with a 30 mL internal volume, accommodates samples up to 20 mg. Sealing of the larger capsule should be carried out in the presence of helium or argon to prevent entrapment of air. Requires but does not include Tin Capsules either P/N N2411255, P/N N2411362, or P/N N2410155.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.