Learn More
Glas Col LLC Heating Mantle
Supplier: Glas Col LLC 100B TM106
Aluminum housed heating mantle 500ml round bottom flask to 400 degrees C maximum internal operating temperature.115V, 270W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. . All 115 volt units are CSA certified.Series TM mantles are available with a bottom hole for flask drains.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.