Learn More
NSI Environmental Solutions GGA CONCENTRATE
Supplier: NSI Environmental Solutions QCI081SNIP
GGa Concentrate Snips for BOD5. a box of 25 single-use snips, each containing 6 mL of Glucose: Glutamic acid solution (300 mg/L total). Just snip open the tube and pour contents directly into a 300 mL BOD bottle. Yields a 2% solution of GGa as per SM 5210B. We've sterilized the solution in the snip, it is sterile indefinitely at room temperature until opened.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.