Learn More
Restek™ GC-MS Cleaning Kit
Signs that GC-MS instruments may need cleaning include poor sensitivity, loss of sensitivity at high masses, and high multiplier gain during an auto tune. This kit includes all of the necessary components for cleaning and polishing ion sources.
$327.00 - $432.00
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
06-611-095
|
Restek™
27194 |
With Dremel Tool |
Each for $432.00
|
|
|||||
06-611-096
|
Restek™
27195 |
Without Dremel Tool |
Each for $327.00
|
|
|||||
Description
- Cleaning Kits are available either with or without battery powered Dremel tool
Specifications
GC-MS Cleaning Kit | |
1 Kit |