Learn More
Enzo Life Sciences SCREEN-WELL GABAergic ligand library (100µl/well)
Supplier: Enzo Life Sciences BML28190100
The GABAergic Ligands library contains 56 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The GABAergics Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 CNS receptor ligands. Long Term Storage: -80°C. Applications: HTS .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.