Learn More
New England Biolabs, Inc. G(5')ppp(5') RNA Cap Structure Analog – 5 umol
Supplier: New England Biolabs, Inc. S1407L
The 5' terminal m7G cap present on most eukaryotic mRNAs promotes translation in vitro at the initiation level. For most RNAs, elimination of the cap structure causes a loss of stability, especially against exonuclease degradation, and a decrease in the formation of the initiation complex of mRNAs for protein synthesis. Certain prokaryotic mRNAs containing a 5' terminal cap structure are translated as efficiently as or more efficiently than eukaryotic mRNAs in a eukaryotic cell-free protein synthesizing system. Also a cap requirement has been observed for splicing eukaryotic substrate RNAs. A method using E. coli RNA Polymerase primed with m7G(5' )ppp(5' )G or m7G(5' )ppp(5' )A for an efficient in vitro synthesis of capped RNAs has been developed by Contreas. Larger amounts of capped RNAs are produced by transcription systems using SP6 RNA polymerase primed with m7G(5' )ppp(5' )G).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.