Learn More
Perkin Elmer US LLC FURA 2 Excitation Filter Set, 340/330 nm or 340/365 nm
Supplier:  Perkin Elmer US LLC L2250021
Encompass_Preferred
If using FURA 2 and BCECF, which are both excitation dyes, one drive unit (P/N L2250146) is required to fit on both pairs of filters. Both dyes can be monitored in the same experiment. If using INDO-1 and BCECF, two drive units are necessary- one for excitation and one for emission.
Catalog No. 50-211-2674