Learn More
Yamato Scientific America, Inc. Yamato 150L Forced and Natural Convection Oven, Energy saving, 220V
Supplier: Yamato Scientific America, Inc. DNF611
High-performance, energy-saving, programmable 150L constant temperature oven with two types of circulation (forced and natural convection) in one unit. Program featured to reduce power consumption significantly. Superior heat tightness and insulation of chamber. Excellent dust tightness, dust can hardly enter the chamber. Air velocity changeable in 10 stages using digital setting of controller. Standard with 99 step program operation with repeat operation, auto start, auto stop and quick auto stop functions. Adjustable damper position at chamber front to optimize operation. Fluorescent display, interactive input method, calibration off-set function. Operating temp ambient +15~260°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.