Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Digital Paddlewheel Flowmeter, 0.08 to 10 GPM, PP, 4-20mA, w/ RTD
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3275536
Designed for water-based liquids, these IP65-rated flowmeters provide flow measurement to 35 GPM (132.5 LPM) and models with the RTD option measure temperature to 140¿F (60¿C). The RTD is platinum encased in 316 stainless steel and has a 4-wire 12" (30.5 cm) cable for output. Flow indication can be seen via acrylic cover. Mount units horizontally or vertically.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.