Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter with Valve, Direct Reading, PTFE Fittings, 65-mm Scale; 20 std mL/min Carbon Dioxide
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3200608
These meters eliminate the need for correlation charts with direct-reading scales for individual routine gases. The inert PTFE and glass wetted parts combine with the rigid anodized aluminum frame to offer a unique solution for ultra-pure gas applications. Flowtubes are replaceable.