Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area In-Line Flowmeter with Valve, Direct Reading, PTFE and PCTEF Fittings, 3/8"" NPT(F); 2,000 ml/min Water
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3205710
Ideal for use in applications requiring the high purity/high temperature suitability that fluoropolymers are capable of. Common applications include those with acids, caustics, solvents, and high purity liquids. The solid PTFE float is designed to maximize each meter's flow range so that many of the available models provide an outstanding turn-down ratio. Flowmeters have direct-reading water scales. *Leak integrity rating of 1 x 10-7 sccs Helium or better.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.