Learn More
Perkin Elmer US LLC Flowcell for Polarimetry
Supplier:  Perkin Elmer US LLC B0017054
Encompass_Preferred
PerkinElmer offers a wide range of polarimetry cells for virtually all applications, including micro cells, short path cells, flow cells, and fused silica (quartz) cells. Quartz cells are designed for elevated temperatures ( 200° max) and can be used for all wavelengths. Glass cells are designed for visible wavelengths above 400 nm.
Catalog No. 50-904-2085