Learn More
Ace Glass, Inc. Flask, reaction, 30L, 200mm(8in) ground Duran; O-ring groove flange, flush-seal bottom valve
Supplier: Ace Glass, Inc. 652283
30L cylindrical reaction flask with Duran flat-ground 200mm(8) o-ring groove flange, and a flush-seal bottom valve. For use with a stainless steel quick release clamp. The flask is supplied with a CAPFE o-ring.