Learn More
Fisherbrand™ Tubing Four-Way Polypropylene Connectors
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Work in any tubing or vacuum system.
This item is not returnable.
View return policy
Description
- Excellent chemical and temperature resistance
- Serrations hold tubing tightly and prevent slipping
Specifications
4-Way Connector | |
0.18 to 0.21 in. | |
1.39 in. | |
4.5 to 5.3mm | |
5.8 mm | |
4-way | |
0.136 in. | |
20 psi |
Autoclavable | |
Polypropylene | |
35.3 mm | |
Used for any tubing or vacuum system | |
0.23 in. | |
3.45 mm | |
Transparent |