Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Bovine Serum Albumin (BSA) Low Endotoxin Powder
Purified by a heat shock process with additional treatments
Description
CAS: 9048-46-8
Specifications
BSA Low Endotoxin | |
Proprietary heat shock process | |
Bovine |
6.5 to 7.5 | |
100g | |
Powder |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction