Learn More
Essendant True HEPA Filter for Fellowes 290 Air Purifiers, 12.63 x 16.31
Supplier:  Essendant FEL9287201
Encompass_Preferred
True HEPA filter captures 99.97% of airborne particles as small as 0.3 microns, including pollen, ragweed and other allergens, viruses, germs, dust mites, mold spores and odors. AeraSafe™ antimicrobial treatment provides built-in protection from the growth of odor-causing bacteria, mildew and fungi on the filter. Compatible with AeraMax Air Purifiers. Use in the office, home office, school, or wherever you need to filter the air around you.
Catalog No. 27-903-22