Learn More
Sigma Aldrich Fine Chemicals Biosciences Estuarine Sediment Reference Material | Certified Elemental Composition: Al, As, Cd, Cr, Cu, Fe, Hg, Mn, Ni, Pb, V, Zn | NIST SRM™ 1944 | 70 GR
Supplier: Sigma Aldrich Fine Chemicals Biosciences NIST1646A
Estuarine sediment reference material | Elemental composition: Al, As, Cd, Cr, Cu, Fe, Hg, Mn, Ni, Pb, V, Zn certified | CAS No: N/A | NIST SRM 1944 | 70g