Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ OptiScale™ Capsule Filters for Aseptic Processing Applications
Small-volume option for process screening and scaling
Supplier: MilliporeSigma™ SHGLA47FF3
Description
- With “drop-in” filters
- Non-sterile
- PVDF vent cap
- Wettability: Hydrophilic
- Housing and drainage disk: Polycarbonate
- Filter support: Polypropylene
- Filtration Area: 17.7cm2
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
Specifications
0.45/0.22 μm | |
17.7 cm2 | |
Polycarbonate | |
7.4 cm |
Non-sterile | |
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates | |
2.91 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction