Learn More
Biomatik Corporation Phospho-mTOR (Ser2448) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51512
Encompass_Preferred
Phospho-mTOR (Ser2448) Colorimetric Cell-Based ELISA Kit for detecting Phospho-mTOR (Ser2448) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42345. Gene ID: 2475. Target Synonyms: FRAP; FRAP1; FRAP2; RAFT1; Rapamycin target protein; kinase mTOR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9071