Learn More
Bioanalytical Systems Inc EC EPSILON METHODS SFTWR
Supplier: Bioanalytical Systems Inc EF1677
Upgrade, Software; BASi; Epsilon - EC Methods; Upgrading for allowed techniques; Sequential techniques strings together up to 6 techniques and repeat the cycle up to 999 times