Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ebselen
Glutathione peroxidase mimic; peroxynitrite scavenger
$122.96 - $512.72
Specifications
Purity | 98% |
---|---|
Molecular Formula | 2-Phenyl-1,2-benzisoselenazol-3(2 H)-one |
CAS | 60940-34-3 |
Content And Storage | -20°C |
Product Type | Ebselen |
Description
Glutathione peroxidase mimic; peroxynitrite scavenger. Inhibits ferroptosis. Also inhibits lipoxygenase, cyclooxygenase, nitric oxide synthase, protein kinase C and H+/K+-ATPase activity. Inhibits the hepatic carcinogenic effects of aflatoxin B1. Antioxidant and anti-inflammatory. Selectively inhibits Gq protein signaling and enhances differentiation of brown adipocytes. Inhibits SARS-CoV-2 Mpro in vitro (IC50 = 0.67 μM in FRET assay)Specifications
98% | |
60940-34-3 | |
Ebselen | |
Ebselen |
2-Phenyl-1,2-benzisoselenazol-3(2 H)-one | |
-20°C | |
M.W. (Observed): 274.18 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title