Learn More
New England Biolabs, Inc. E.coli Poly (A) Polymerase – 500 units
Supplier: New England Biolabs, Inc. M0276L
Poly(A) Polymerase catalyzes the template independent addition of AMP from ATP to the 3 end of RNA.