Learn More
Waters Corp Flowcell, Dummy, 2465
Supplier:  Waters Corp 700001943
Encompass_Preferred
The 2465 Dummy Cell consists of a resistor (R) and a capacitor (C) in parallel. A successful dummy cell test confirms that the controller, including the cell cable, functions properly.
Catalog No. 50-792-647