Learn More
Yamato Scientific America, Inc. Yamato 300L Dry Sterilizer, Forced Convection, 220V
Supplier: Yamato Scientific America, Inc. SK811
Programmable forcedl convection 300L dry heat sterilizer with independent overheat prevention device. High precision controller with improved display visibility and operability. Standard equipped with calibration off-set, lock function, power recovery mode, power on and operation time accumulation, calendar time, accumulation power consumption monitoring, total CO₂ emission, and heat output, save and access operator setting information. Maximum 99 steps, 99 patterns, repeat operation. Easy sample data collection with cable port. Temp setting range ambient +10~210°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.