Learn More
Seca Corp 856 Digital organ and diaper scale with stainless steel cover
SDP
Supplier:  Seca Corp 8561314009
Encompass_Preferred
The diaper/organ scale seca 856 has been designed to meet the specific requirements in its area of application. The raised display is protected from soiling by fluids and the specially sealed battery compartment keeps moisture away from the electronic parts. This scale also fulfills tough hygienic standards with a sealed stainless steel surface thatʼs easy to clean. With the aid of the pre-TARE function, the additional weight of containers can simply be deducted for precise determination of net weight. The fine 1-gram graduation ensures highly precise results. • Complete stainless steel-surface reduces contamination • Easy to clean and disinfect • Scale completely sealed from moisture • Pre-TARE function for permanent use with the container of your choice • Increase in metal housing protects display from liquid • Capacity: 11 lbs / 5 kg • Graduation: 0.05 oz < 6.6 lbs > 0.1 oz / 1 g < 3 kg > 2 g • Functions: TARE, pre-TARE, HOLD, lbs/kg switch-over, automatic switch-off
Catalog No. NC1798792