Learn More
Medchemexpress LLC HY-110281 50mg Medchemexpress, Dehydroascorbic acid CAS:490-83-5 Purity:>98%
Supplier: Medchemexpress LLC HY11028150MG
Medchemexpress, HY-110281 50mg Dehydroascorbic acid CAS:490-83-5 Dehydroascorbic acid, a blood-brain barrier transportable form of vitamin C, mediates potent cerebroprotection in experimental stroke. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.